Learn More
Fluorescamine, Thermo Scientific Chemicals
For fluorescent detection of amino acids, peptides and proteins
Supplier: Thermo Scientific Chemicals 043749ME
Description
For fluorescent detection of amino acids, peptides and proteins.Fluorescamine plays an important role as a reagent for the detection of amines, peptides and proteins. It is also used as a substrate for measuring protease activity. It is utilized in the detection and quantitation of residual aminopenicillins and sulfonamides in honey by HPLC. It finds application in the determination of lisinopril in human plasma and urine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluorescamine | |
155°C to 158°C | |
C17H10O4 | |
500 mg | |
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion | |
ZFKJVJIDPQDDFY-UHFFFAOYNA-N | |
4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione | |
37927 | |
Powder |
38183-12-9 | |
White to Cream | |
MFCD00005928 | |
14,4158 | |
Soluble in acetone,ethanol,chloroform,dimethylsulfoxide and acetonitrile. Slightly soluble in water. | |
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12 | |
278.26 | |
278.27 |
Chemical Identifiers
38183-12-9 | |
278.26 | |
ZFKJVJIDPQDDFY-UHFFFAOYNA-N | |
37927 |
C17H10O4 | |
MFCD00005928 | |
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion | |
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12 |
Safety and Handling
EINECSNumber : 253-814-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only