missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular Probes™ FluoReporter™ Mini-Biotin-XX Protein Labeling Kit
FluoReporter™ Mini-Biotin-XX Protein Labeling Kit
Manufacturer: Molecular Probes™ F6347
Description
The FluoReporter™ Mini-biotin-XX Protein Labeling Kit provides a convenient and flexible means for attaching biotin-XX to your antibody or protein (>30 kDa). This kit contains a water-soluble biotin-XX sulfosuccinimidyl ester, where “XX” is a 14-atom spacer that separates the reactive ester from the biotin moiety, making the biotin more accessible to avidin or streptavidin. The kit supplies the reagents needed for 5 to 10 labeling and separation reactions.Important Features of FluoReporter™ Protein Labeling Kits:
Learn More About Protein Labeling
We offer a wide selection of Molecular Probes™ protein and antibody labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.
We’ll Make a Custom Labeled Protein for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom labeled protein for you. Our custom services are efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Protein Labeling Kit | |
Biotin-Based | |
Room Temperature | |
Biotin | |
Antibodies (General), Proteins (General) |
Store in refrigerator at 2°C to 8°C | |
0.1 to 3 mg | |
FluoReporter™ | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.