missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular Probes™ FluoReporter™ FITC Protein Labeling Kit
FluoReporter™ FITC Protein Labeling Kit
Manufacturer: Molecular Probes™ F6434
Description
The FluoReporter™ FITC Protein Labeling Kit provides a convenient and flexible means for attaching fluorescein (FITC, Ex/Em ∼494/518 nm) to your monoclonal or polyclonal antibody (or any protein >30 kDa). The kit supplies the reagents needed for 5 to 10 labeling and separation reactions.Important Features of the FluoReporter™ FITC Protein Labeling Kit:
Learn More About Protein Labeling
We offer a wide selection of Molecular Probes™ protein and antibody labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.
We’ll Make a Custom Labeled Protein for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom labeled protein for you. Our custom services are efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Protein Labeling Kit | |
Fluorescent | |
Room Temperature | |
FITC (Fluorescein) |
Store in refrigerator 2°C to 8°C and protect from light. | |
0.2 to 2 mg | |
FluoReporter™ | |
Antibodies (General), Proteins (General) |
For Research Use Only. Not for use in diagnostic procedures.