missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluorene-4-carboxylic acid, 96%

$83.27 - $721.72

Chemical Identifiers

CAS 6954-55-8
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
MDL Number MFCD00001112
InChI Key WJNBLUOXTBTGMB-UHFFFAOYSA-N
Synonym fluorene-4-carboxylic acid, 4-fluorenecarboxylic acid, maybridge1_002107, 9h-fluorene-4-carboxylicacid, fluorene-4-carboxylic acid, technical grade
PubChem CID 81402
IUPAC Name 9H-fluorene-4-carboxylic acid
SMILES C1C2=CC=CC=C2C3=C(C=CC=C31)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL01164MD View Documents Alfa Aesar™
L01164MD
250mg Each for $83.27
 
AAL0116403 View Documents Alfa Aesar™
L0116403
1g Each for $222.07
 
AAL0116406 View Documents Alfa Aesar™
L0116406
5g Each for $721.72
 
Description

Description

Applications
Fluorene-4-carboxylic acid is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

6954-55-8
210.232
WJNBLUOXTBTGMB-UHFFFAOYSA-N
81402
C1C2=CC=CC=C2C3=C(C=CC=C31)C(=O)O
C14H10O2
MFCD00001112
fluorene-4-carboxylic acid, 4-fluorenecarboxylic acid, maybridge1_002107, 9h-fluorene-4-carboxylicacid, fluorene-4-carboxylic acid, technical grade
9H-fluorene-4-carboxylic acid

Specifications

191°C to 192°C
6954-55-8
MFCD00001112
fluorene-4-carboxylic acid, 4-fluorenecarboxylic acid, maybridge1_002107, 9h-fluorene-4-carboxylicacid, fluorene-4-carboxylic acid, technical grade
C1C2=CC=CC=C2C3=C(C=CC=C31)C(=O)O
210.232
210.23
Fluorene-4-carboxylic acid
250mg
C14H10O2
2050637
WJNBLUOXTBTGMB-UHFFFAOYSA-N
9H-fluorene-4-carboxylic acid
81402
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning