missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluorene-2-carboxaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1491106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluorene-2-carboxaldehyde | |
82°C to 86°C | |
MFCD00001123 | |
2255852 | |
2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, # | |
O=CC1=CC=C2C(CC3=CC=CC=C23)=C1 | |
194.23 | |
194.23 |
30084-90-3 | |
C14H10O | |
5 g | |
Air Sensitive | |
MNQGEQSXFDKAPY-UHFFFAOYSA-N | |
9H-fluorene-2-carbaldehyde | |
34804 | |
99% |
Chemical Identifiers
30084-90-3 | |
194.23 | |
MNQGEQSXFDKAPY-UHFFFAOYSA-N | |
34804 | |
O=CC1=CC=C2C(CC3=CC=CC=C23)=C1 |
C14H10O | |
MFCD00001123 | |
2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, # | |
9H-fluorene-2-carbaldehyde |
Safety and Handling
H500
EINECSNumber : 250-035-2
RTECSNumber : LL5917000
TSCA : No
RUO – Research Use Only