Please login to your online account to display your discounted pricing

Alfa Aesar™ Fluorene-1-carboxylic acid, 97%

$123.77 - $2,099.58

Chemical Identifiers

Chemical Name or Material Fluorene-1-carboxylic acid
CAS 6276-03-5
Molecular Formula C14H10O2
Formula Weight 210.23
MDL Number MFCD00001130
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2502003

 
alfa aesar™
B2502003
1g Each for $123.77

AAB2502006

 
alfa aesar™
B2502006
5g Each for $511.04

AAB2502014

 
alfa aesar™
B2502014
25g Each for $2,099.58
Description & Specifications

Specifications

Formula Weight 210.23
Molecular Formula C14H10O2
CAS 6276-03-5
Melting Point 246° to 248°C
Chemical Name or Material Fluorene-1-carboxylic acid
Assay 97%
MDL Number MFCD00001130
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.