missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoranthene 98.0+%, TCI America™
Supplier: TCI America F0016500G
Specifications
Fluoranthene | |
110°C | |
384°C | |
MFCD00001184 | |
idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 | |
202.26 | |
CHEBI:33083 | |
≥98.0% (GC) |
206-44-0 | |
Yellow | |
C16H10 | |
500 g | |
GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
fluoranthene | |
9154 | |
202.26 | |
Crystalline Powder |
Chemical Identifiers
206-44-0 | |
202.26 | |
GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
9154 | |
fluoranthene |
C16H10 | |
MFCD00001184 | |
idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
CHEBI:33083 | |
C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 |
Safety and Handling
EINECSNumber : (4)-0002
RTECSNumber : LL4025000
TSCA : Yes