Please login to your online account to display your discounted pricing

Fluocinolone Acetonide, 98.92%, For HPLC analysis, MP Biomedicals™


Soluble in methanol, ethanol, chloroform and actone; slightly soluble in ether; practically insoluble in water

Quantity 1g

Manufacturer: mp biomedicals  0215113191

 View more versions of this product

Catalog No. ICN15113191

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
Chemical Name or Material Fluocinolone Acetonide
Name Note For HPLC analysis
CAS 67-73-2
Physical Form Free Flowing White Powder
Molecular Formula C24H30F2O6
Formula Weight 452.5g/mol
MDL Number MFCD00010525
Synonym Sinalar
EINECS Number 200-668-5

Specifications

Chemical Name or Material Fluocinolone Acetonide
Name Note For HPLC analysis
CAS 67-73-2
Physical Form Free Flowing White Powder
Molecular Formula C24H30F2O6
Formula Weight 452.5g/mol
MDL Number MFCD00010525
Synonym Sinalar