missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluocinolone Acetonide, 98.92%, For HPLC analysis, MP Biomedicals™

Soluble in methanol, ethanol, chloroform and actone; slightly soluble in ether; practically insoluble in water

$108.19 - $228.60

Chemical Identifiers

CAS 67-73-2
Physical Form Free-Flowing Powder
Molecular Formula C24H30F2O6
Formula Weight 452.5
MDL Number MFCD00010525
Synonym Sinalar
Chemical Name or Material Fluocinolone Acetonide, For HPLC analysis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15113183 Mp Biomedicals
15113183
250mg Each for $228.60
ICN15113180 Mp Biomedicals
15113180
100mg Each for $108.19
Specifications Description & Specifications

Chemical Identifiers

67-73-2
C24H30F2O6
MFCD00010525
Fluocinolone Acetonide, For HPLC analysis
Free-Flowing Powder
452.5
Sinalar

Specifications

White
67-73-2
C24H30F2O6
MFCD00010525
Fluocinolone Acetonide, For HPLC analysis
250mg
Free-Flowing Powder
452.5
Sinalar

Safety and Handling

EINECSNumber : 200-668-5