Please login to your online account to display your discounted pricing

Flumazenil, Tocris


Benzodiazepine antagonist

Quantity 10mg

Manufacturer: r & d systems  132810

 View more versions of this product

Catalog No. 132810

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 303.29
Quantity 10mg
CAS 78755-81-4
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Synonym 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, ethyl ester
Molecular Formula C15H14FN3O3
Chemical Name or Material Flumazenil

Specifications

Chemical Name or Material Flumazenil
Assay Percent Range >99%
Molecular Formula C15H14FN3O3
Formula Weight 303.29
Synonym 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, ethyl ester
Accessories

Desiccate at +4°C