Please login to your online account to display your discounted pricing

Flumazenil, Tocris

Benzodiazepine antagonist

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material Flumazenil
Assay Percent Range >99%
Molecular Formula C15H14FN3O3
Formula Weight 303.29
Synonym 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

132850

 
r & d systems
132850
50mg Each for $478.51

132810

 
r & d systems
132810
10mg Each for $113.57
Description & Specifications

Specifications

Formula Weight 303.29
Quantity 50mg
Assay Percent Range >99%
CAS 78755-81-4
Recommended Storage Desiccate at +4°C
Chemical Name or Material Flumazenil
Molecular Formula C15H14FN3O3
Synonym 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, ethyl ester
Safety and Handling

Desiccate at +4°C