missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Flumazenil, 98%, Thermo Scientific Chemicals

Benzodiazepine antagonist

Supplier:  Thermo Scientific Chemicals J62537MC

 View more versions of this product

Catalog No. AAJ62537MC



Description

Description

Benzodiazepine antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Flumazenil
White
MFCD00242764
14,4135
Soluble in DMSO to 25mM
CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C
303.293
CHEBI:5103
98%
78755-81-4
C15H14FN3O3
100 mg
flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
OFBIFZUFASYYRE-UHFFFAOYSA-N
ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
3373
303.29
Powder

Chemical Identifiers

78755-81-4
303.293
OFBIFZUFASYYRE-UHFFFAOYSA-N
3373
ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
C15H14FN3O3
MFCD00242764
flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
CHEBI:5103
CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C
Safety and Handling

Safety and Handling

RTECSNumber : NI2922170

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only