Please login to your online account to display your discounted pricing

Alfa Aesar™ Flufenamic acid, 97%

Chemical Identifiers

Chemical Name or Material Flufenamic acid
CAS 530-78-9
Molecular Formula C14H10F3NO2
Formula Weight 281.23
MDL Number MFCD00002422
UN Number UN2811
Beilstein 1996069
Merck Index 14,4132
Synonym N-(3-Trifluoromethylphenyl)anthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2358309

  SDS
alfa aesar™
B2358309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2358318

  SDS
alfa aesar™
B2358318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2358330

  SDS
alfa aesar™
B2358330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 281.23
Molecular Formula C14H10F3NO2
Health Hazard 1 Warning
Quantity 10g
Merck Index 14,4132
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
MDL Number MFCD00002422
RTECS Number CB4375000
UN Number UN2811
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Beilstein 1996069
CAS 530-78-9
Synonym N-(3-Trifluoromethylphenyl)anthranilic acid
Melting Point 133° to 135°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Flufenamic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.