FLT3 Recombinant Human Protein DFS Item
FLT3 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV4108
 View more versions of this product
Catalog No. PV4108
Description
Description
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.
Specifications
Specifications
NP_004110
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
Frozen
FLT3 Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
FLT3, FLK2, STK1, CD135
FLT3 was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 2.5mM DTT, 0.01% NP-40, 200μM ATP, 200μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
Amino acids 564-958
2430nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
His Tag
0.35mg/mL
Frozen
48.6kDa
90% by SDS-PAGE
–80°C
1 Each
FLT3 Recombinant Human Protein
Kinase assay
PDGFR
FLT3 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.