missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Human FLT3, His Tag Recombinant Protein
FLT3 Recombinant Human Protein
Supplier: Thermo Fisher Scientific PV3182
Description
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.Specifications
NP_004110 | |
2322 | |
10μg | |
PDGFR Kinase Family | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
FLT3 | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
48.6 kDa | |
FLT3, FLK2, STK1, CD135 | |
aa 564-958 | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
fms-related tyrosine kinase 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.