FLT3 [D835Y] Recombinant Human Protein, Invitrogen™ DFS Item
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family.
Manufacturer:  Life Technologies LSPV4271
 View more versions of this product
Catalog No. PV4271
Description
Description
The FLT3 D835Y mutant is a drug resistant form of FLT3.
Specifications
Specifications
NP_004110, D835Y
Frozen
48.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
PDGFR Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 100mM NaCl, 0.5mM EDTA, 0.05% NP-40, 2mM DTT, 50% glycerol
1mg
RUO - research use only
FLT3 D835Y, FLK2, STK1, CD135
Baculovirus
Human
His Tag
Documents