FLT1 (VEGFR1) Recombinant Human Protein DFS Item
FLT1 (VEGFR1) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3666
 View more versions of this product
Catalog No. PV3666
Description
Description
FLT1 (VEGFR1) is a receptor tyrosine kinase which plays a critical role in angiogenesis. Active angiogenesis has been found to play important role in many pathological and physiological processes, such as tumor progression, diabetic retinopathy, rheumatoid arthritis and wound healing.
Specifications
Specifications
NP_002010
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation
92.6kDa
90% by SDS-PAGE
–80°C
1 Each
FLT1 (VEGFR1), VEGFR1, FLT
FLT1 (VEGFR1) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β−glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinases and inhibitors
VEGF receptors
FLT1 (VEGFR1) Recombinant Human Protein
Human
0.42mg/mL
2321
FLT1 (VEGFR1) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Receptor tyrosine kinases
FLT1 (VEGFR1) Recombinant Human Protein
Kinase assay
Baculovirus
aa 781 to 1338
520nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.