Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Flt-1/VEGFR1 Ab-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ Flt-1/VEGFR1 Ab-1, Rabbit Polyclonal Antibody.

$271.02 - $1,379.02

Specifications

Antigen Flt-1/VEGFR1 Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1527P

 
thermo scientific™ lab vision™
RB1527P
Purified Ab with BSA and Azide 1mL Each for $1,260.14

RB1527P0

 
thermo scientific™ lab vision™
RB1527P0
Purified Ab with BSA and Azide 100µL Each for $271.02

RB1527P1

 
thermo scientific™ lab vision™
RB1527P1
Purified Ab with BSA and Azide 500µL Each for $762.53

RB1527P1ABX

 
thermo scientific™ lab vision™
RB1527P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $838.59

RB1527PABX

 
thermo scientific™ lab vision™
RB1527PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,379.02
Description & Specifications

Specifications

Antigen Flt-1/VEGFR1 Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Angiogenesis
Type Primary
Immunogen A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin
Isotype IgG

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ Flt-1/VEGFR1 Ab-1, Rabbit Polyclonal Antibody.

Flt-1/vascular endothelial growth factor receptor 1 (VEGFR1), a cell membrane receptor kinase, is a high-affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flk-1 and Flt-4.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the Flt-1 precursor of human origin

Molecular Weight: 180 kDa

Positive Control: Hey cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO