Invitrogen™ eBioscience™ Flow Cytometry Staining Buffer
Designed for use in immunofluorescent staining protocols of cells in suspension. Flow Cytometry Staining Buffer is a buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative.
Manufacturer:  Invitrogen™ LS00422226
 View more versions of this product
Catalog No. 501129748
Alert:
Contains Sodium azide
Description
Description
  • This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis
  • Buffer contains animal serum proteins to help minimizing non-specific binding of antibodies
  • Use within six months of opening
Specifications
Specifications
Flow Cytometric Analysis
2° to 8°C.
600mL