Please login to your online account to display your discounted pricing

Florisil™, 60 to 100 Mesh, J.T.Baker™


Selective Adsorbent for Gas Chromatography

Quantity 2kg
Packaging Poly Bottle

Manufacturer: avantor performance  M36808

 View more versions of this product

Catalog No. 02003118

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Florisil™
Mesh Size 60 to 100 Mesh
Packaging Poly Bottle
CAS 1343-88-0
Specific Gravity 1L = 2.51kg
Quantity 2kg

Specifications

Chemical Name or Material Florisil™
CAS 1343-88-0