missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Florisil, 60-100 mesh, for column chromatography, Thermo Scientific Chemicals
Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle
Supplier: Thermo Scientific Chemicals 205455000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatographySpecifications
Fine Powder | |
Florisil™ | |
1343-88-0 | |
MgO3Si | |
01,393; 09,223 | |
Glass Bottle | |
ZADYMNAVLSWLEQ-UHFFFAOYSA-N | |
100.39 |
White | |
60-100 mesh, for column chromatography | |
500 g | |
MFCD00078249 | |
10, 5509 | |
Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9% | |
100.39 |
Chemical Identifiers
1343-88-0 | |
100.39 | |
ZADYMNAVLSWLEQ-UHFFFAOYSA-N |
MgO3Si | |
MFCD00078249 |
RUO – Research Use Only