Please login to your online account to display your discounted pricing

Florisil, 60-100 mesh, for column chromatography, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: fisher scientific acros  205452500

 View more versions of this product

Catalog No. AC205452500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Fine Powder
Formula Weight 100.39
Quantity 250g
Color White to off-white
Fieser 01,393; 09,223
Merck Index 10, 5509
Packaging Glass bottle
Chemical Name or Material Florisil™
Molecular Formula MgO3Si
MDL Number MFCD00078249
Name Note 60-100 mesh, for column chromatography
Particle Size Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9%
CAS 1343-88-0

Specifications

Chemical Name or Material Florisil™
Name Note 60-100 mesh, for column chromatography
CAS 1343-88-0
Molecular Formula MgO3Si
Formula Weight 100.39
MDL Number MFCD00078249