missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Florfenicol, 98%

Manufacturer:  Alfa Aesar™ J6699506

 View more versions of this product

Catalog No. AAJ6699506



Description

Description

  • A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
Specifications

Specifications

Florfenicol
C12H14Cl2FNO4S
14,4109
AYIRNRDRBQJXIF-NXEZZACHSA-N
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
114811
358.21
98%
White
5g
73231-34-2
MFCD00864834
Soluble in ethanol to 25mM and in DMSO to 100mM
CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
358.205
CHEBI:87185
Powder
618°C
152°C to 156°C
>110°C (230°F)

Chemical Identifiers

73231-34-2
358.205
AYIRNRDRBQJXIF-NXEZZACHSA-N
CHEBI:87185
CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
C12H14Cl2FNO4S
MFCD00864834
114811
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
Safety and Handling

Safety and Handling

RTECSNumber : DB6034000

TSCA : No

Recommended Storage : Ambient temperatures

Documents