missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Flk-1/KDR/VEGFR2 Ab-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Flk-1/KDR/VEGFR2 Ab-1, Rabbit Polyclonal Antibody.

$296.45 - $1,477.33

Specifications

Antigen Flk-1/KDR/VEGFR2 Ab-1
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human,Mouse,Human,Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1526P Thermo Scientific™ Lab Vision™
RB1526P
Purified Ab with BSA and Azide 1mL Each for $1,377.08
RB1526P0 Thermo Scientific™ Lab Vision™
RB1526P0
Purified Ab with BSA and Azide 100μL Each for $296.45
RB1526P1 Thermo Scientific™ Lab Vision™
RB1526P1
Purified Ab with BSA and Azide 500μL Each for $832.57
RB1526P1ABX Thermo Scientific™ Lab Vision™
RB1526P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $915.71
RB1526PABX Thermo Scientific™ Lab Vision™
RB1526PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,477.33
RB1526R7 Thermo Scientific™ Lab Vision™
RB1526R7
Ready-to-use for IHC (UVLP) 7mL Each for $720.55
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Flk-1/KDR/VEGFR2 Ab-1, Rabbit Polyclonal Antibody.

Flk-1/KDR vascular endothelial growth factor receptor 2, a cell membrane receptor kinase, is a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flt-1 and Flt-4.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of the precursor form of the mouse Flk-1

Molecular Weight: ∽180 kDa

Positive Control: Hey cells, Angiosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Flk-1/KDR/VEGFR2 Ab-1
Polyclonal
Human,Mouse,Human,Mouse
RUO
Primary
IgG
Western Blot
Unlabeled
Rabbit
Angiogenesis
A synthetic peptide from C-terminus of the precursor form of the mouse Flk-1