Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Flk-1/KDR/VEGFR2 Ab-1, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Flk-1/KDR/VEGFR2 Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1526P1ABX

 View more versions of this product

Catalog No. RB1526P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Applications Western Blotting
Immunogen A synthetic peptide from C-terminus of the precursor form of the mouse Flk-1
Isotype IgG
Format Concentrated
Host Species Rabbit
Quantity 500µL
Antigen Flk-1/KDR/VEGFR2 Ab-1
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Angiogenesis
Classification Polyclonal
Type Primary
Concentration 1mg/mL
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Flk-1/KDR/VEGFR2 Ab-1, Rabbit Polyclonal Antibody.

Flk-1/KDR vascular endothelial growth factor receptor 2, a cell membrane receptor kinase, is a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin-like sequences in the extracellular domain and exhibits sequence similarity to Flt-1 and Flt-4.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of the precursor form of the mouse Flk-1

Molecular Weight: ∽180 kDa

Positive Control: Hey cells, Angiosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO