missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Flavin Adenine Dinucleotide Disodium Salt Hydrate, TCI America™

Manufacturer:  TCI America F0014100MG

 View more versions of this product

Catalog No. F0014100MG



Specifications

Specifications

Flavin Adenine Dinucleotide Disodium Salt Hydrate
100mg
C27H33N9Na2O16P2
flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
CC1=CC2=C(C=C1C)N(C3=NC(=O)N=C(C3=N2)[O-])CC(C(C(COP(=O)([O-])OP(=O)(O)OCC4C(C(C(O4)N5C=NC6=C5N=CN=C6N)O)O)O)O)O.O.[Na+].[Na+]
847.535
829.52
≥94.0% (HPLC)
Undesignated
84366-81-4
MFCD00041835
GXTPHHZYFMAGLX-UJXBNFGUSA-L
disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
131675332
Crystal-Powder at 20°C

Chemical Identifiers

84366-81-4
847.535
GXTPHHZYFMAGLX-UJXBNFGUSA-L
131675332
CC1=CC2=C(C=C1C)N(C3=NC(=O)N=C(C3=N2)[O-])CC(C(C(COP(=O)([O-])OP(=O)(O)OCC4C(C(C(O4)N5C=NC6=C5N=CN=C6N)O)O)O)O)O.O.[Na+].[Na+]
C27H33N9Na2O16P2
MFCD00041835
flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
Safety and Handling

Safety and Handling

RTECSNumber : AU7470000

TSCA : Yes

Recommended Storage : Refrigerator

Documents