Please login to your online account to display your discounted pricing

Flavanone 98%, Acros Organics


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  146620100

 View more versions of this product

Catalog No. AC146620100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 224.26
Quantity 10g
Color Very slightly yellow
EINECS Number 207-654-8
TSCA TSCA
RTECS Number LK6906500
Melting Point 75° to 78°C
Beilstein 17, 364
Packaging Glass bottle
Synonym 2, 3-Dihydroflavone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Flavanone
Name Note 98%
Assay Percent Range 98%
Molecular Formula C15H12O2
MDL Number MFCD00006841
CAS 487-26-3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Flavanone
Name Note 98%
CAS 487-26-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H12O2
Formula Weight 224.26
MDL Number MFCD00006841
Synonym 2, 3-Dihydroflavone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents