missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Humboldt Flatjaw Pinchcocks

One-piece rust resitant brass clamp

Manufacturer:  Humboldt HUMH8620

 View more versions of this product

Catalog No. 05-850B




Description

Description

One-piece rust resitant brass clamp

  • Nickel-plated
  • Clamp flexible tubing up to 0.62 in. (16mm)

Specifications

Specifications

3.25 in.
Large
Mohr Pinchcock
8.25 cm
Nickel, Steel
SDS
Documents