missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BUCHI FlashPure Select C18

FlashPure Select C18 cartridges are designed for ultimate and unmatched performance. C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase.

Manufacturer:  BUCHI 140000125

 View more versions of this product

Catalog No. 05410091



Description

Description

  • 30 μm spherical particles
  • Functionalized silica cartridges can be reused multiple times
  • Large selection of cartridge sizes from 4g-330g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications

Specifications

C18
Any Flash System
Polypropylene
80 g
Non-Polar Compounds, Ideal for Peptides
2 Pc.
20-35 µm
C18
Documents