BUCHI FlashPure Select Flash Chromatography Cartridges: C18
FlashPure Select cartridges are designed for ultimate and unmatched performance.
Supplier: BUCHI 140000127
Description
- Spherical particles
- 13-17 μm silica, 20-35 μm C18
- Functionalized silica cartridges can be reused multiple times
- Large selection of cartridge sizes from 4 g - 3,000 g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Polypropylene | |
1 Pc. | |
220 g | |
Non-Polar Compounds, Ideal for Peptides |
C18 | |
20 to 35 μm | |
C18 | |
Any Flash System |