BUCHI FlashPure ID Diol
FlashPure ID cartridges are degined for Reveleris™ chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system. Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids.
Manufacturer:  BUCHI 140000018
 View more versions of this product
Catalog No. 05410001
Description
Description
  • RFID for optimal convenience on BUCHI Reveleris™ systems
  • 40 µm irregular particle
  • Selection of cartridge sizes from 4g-330g
Specifications
Specifications
Polypropylene
1 Pc.
24 g
Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
Diol
35 to 45 μm
Diol
Reveleris Flash System