BUCHI FlashPure ExoFlex Alumina Acidic
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. EcoFlex Alumina Acidic has the weakest retention compared to basic and neutral alumina.
$222.30 - $2,940.05
Specifications
For Use With (Application) Polar Compounds
For Use With (Equipment) Any Flash System
Material Polypropylene
Type Acidic Alumina
Column Type Alumina Acidic
View More Specs
Products 10
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity  
05410067
View Documents
BUCHI
140000093
1600 g 3 Pc. Pack of 3 Each for $2,453.28
05410063
View Documents
BUCHI
140000089
160 g 10 Pc. Pack of 10 Each for $873.25
05410064
View Documents
BUCHI
140000090
240 g 10 Pc. Pack of 10 Each for $1,436.40
05410060
View Documents
BUCHI
140000086
24 g 18 Pc. Pack of 18 Each for $498.18
05410068
View Documents
BUCHI
140000094
3200 g 2 Pc. Pack of 2 Each for $2,940.05
05410065
View Documents
BUCHI
140000091
440 g 6 Pc. Pack of 6 Each for $1,326.96
05410061
View Documents
BUCHI
140000087
50 g 12 Pc. Pack of 12 Each for $492.47
05410066
View Documents
BUCHI
140000092
660 g 5 Pc. Pack of 5 Each for $1,768.14
05410062
View Documents
BUCHI
140000088
80 g 12 Pc. Pack of 12 Each for $585.96
05410059
View Documents
BUCHI
140000085
8 g 20 Pc. Pack of 20 Each for $222.30
Description
Description
  • 60 μm irregular particles
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 8g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Polar Compounds
Polypropylene
Alumina Acidic
Any Flash System
Acidic Alumina