missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BUCHI FlashPure ExoFlex Alumina Acidic

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. EcoFlex Alumina Acidic has the weakest retention compared to basic and neutral alumina.

Manufacturer:  BUCHI 140000093

 View more versions of this product

Catalog No. 05410067



Description

Description

  • 60 μm irregular particles
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 8g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications

Specifications

Alumina Acidic
Any Flash System
Polypropylene
1600 g
Polar Compounds
3 Pc.
50 to 75 μm
Acidic Alumina
Documents