BUCHI FlashPure Empty Solid Loader
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatogrpahy. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
Manufacturer:  BUCHI 148634552
 View more versions of this product
Catalog No. 05410136
Description
Description
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Specifications
Empty
Reveleris Solid Loading
Polyethylene
Frits
Solid Loading of Insoluble Samples
20 Pc.
20 g
Empty
Documents