BUCHI FlashPure EcoFlex Silica Lok
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. EcoFlex Silica is ideal for basic applications and polar compounds using organic solvents.
$107.61 - $450.95
Specifications
For Use With (Application) Basic Applications, Polar Compounds
For Use With (Equipment) Any Flash System
Material Polypropylene
Type Silica
Column Type Silica
View More Specs
Products 7
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity  
05410019 View Documents BUCHI
140000037
120 g 10 Pc. Pack of 10 Each for $274.35
05410015 View Documents BUCHI
140000033
12 g 18 Pc. Pack of 18 Each for $128.09
05410020 View Documents BUCHI
140000038
220 g 6 Pc. Pack of 6 Each for $287.25
05410021 View Documents BUCHI
140000039
330 g 5 Pc. Pack of 5 Each for $450.95
05410017 View Documents BUCHI
140000035
40 g 12 Pc. Pack of 12 Each for $167.49
05410014 View Documents BUCHI
140000032
4 g 20 Pc. Pack of 20 Each for $107.61
05410018 View Documents BUCHI
140000036
80 g 10 Pc. Pack of 10 Each for $188.72
Description
Description
  • 50 μm irregular particles
  • Most economical solution without sacrificing quality
  • Large selection of cartridge sizes from 4g-3000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Specifications
Basic Applications, Polar Compounds
Polypropylene
Silica
Any Flash System
Silica