missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BUCHI FlashPure EcoFlex C18
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance. C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase.
Manufacturer: BUCHI 140000052
Description
- 50 μm spherical particles
- Functionalized silica cartridges can be reused multiple times
- Large selection of cartridge sizes from 4g-330g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
C18 | |
Any Flash System | |
Polypropylene | |
80 g |
Non-Polar Compounds, Ideal for Peptides | |
2 Pc. | |
40-60 µm | |
C18 |
Promotions
-
Get 5 Silica Cartridge Bundles with Purchase
Buy a BUCHI Pure C-810 Flash Chromatography System, get five Silica Cartridge Bundles at no additional cost.: 01/31/20