MilliporeSigma™ Supelco™ Flash Chromatography Assembly with Standard Column/Ball Joints, Joint Size: SJ 29/15
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z2022901EA
Catalog No. 111008679
Description
Description
Consists of column, flow controller, and pinch clamp. Order solvent reservoir separately.
Specifications
Specifications
813 mm
Flash Chromatography
100 mL
19 mm
Flash Chromatography
SJ 29/15