MilliporeSigma™ Supelco™ Flash Chromatography Assembly with Standard Ball Joints and Fritted Disc (Porosity C), Joint Size: SJ 29/15
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z4168861EA
Catalog No. 111008761
Description
Description
Consists of column, flow controller, and pinch clamp. Order solvent reservoir separately.
Specifications
Specifications
813 mm
Flash Chromatography
100 mL
19 mm
Flash Chromatography
SJ 29/15