Thermo Scientific™ Flammable-Materials Storage Refrigerator/Freezer
Safely store volatile materials with this Flammable-Materials Storage Refrigerator/Freezer. Engineered with no internal electrical components which could trigger an explosion.
$7,662.50 - $7,662.50
Specifications
Certifications/Compliance UL listed meets NFPA 45 and 70
Color White
Control Mechanical Dial Thermostat/Outside on Rear of Unit
Casters 2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Defrost Ref: Automatic Defrost; Frz: Manual Defrost
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Capacity Plug Type Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Capacity Plug Type Price Quantity  
10FCEETSA Thermo Scientific™
10FCEETSA
115 V/60 Hz 10.1 Cubic Feet/286 Liters Yes/Three prong NEMA 5-15P, 3.048 Meters (10 Feet) Each for $7,662.50
Specifications
Description
Safely store volatile materials with the Thermo Scientific™ Flammable-Materials Storage Refrigerator/Freezer. Engineered with no internal electrical components which could trigger an explosion. The refrigerator/freezer meets OSHA and National Fire Protection Association safety requirements.
Features:
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and coolant
  • Three adjustable shelves and four door shelves in refrigerator section
  • Hydraulic thermostat; adjustable natural air flow vent
  • ABS plastic interior with enameled steel exterior
  • Manual defrost
  • Refrigerator/freezer has separate doors
  • Warranty varies by country, please contact us for details
Warranty
13 Months Parts and Labor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
UL listed meets NFPA 45 and 70
Mechanical Dial Thermostat/Outside on Rear of Unit
Ref: Automatic Defrost; Frz: Manual Defrost
2(2 compartments)
Enameled steel
24 x 23.63 x 59.75 in. (60.96 x 60 x 151.77cm)
White
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Bottom Mounted
Hinged, Open Left to Right, Reversible
ABS Plastic
3 Adjustable Shelves, 1 Fixed Shelf, 1 Drawer, 4 Door Shelves