Please login to your online account to display your discounted pricing

FK 3311, Tocris

Cyclooxygenase-2 (COX-2) inhibitor

$113.57 - $239.63

Chemical Identifiers

Chemical Name or Material FK 3311
Assay Percent Range >98%
Molecular Formula C15H13F2NO4S
Formula Weight 341.33
Synonym N-[4-Acetyl-2-(2, 4-difluorophenoxy)phenyl]methanesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

27761

 
r & d systems
RDS27761
1mg Each for $113.57

277610

 
r & d systems
RDS277610
10mg Each for $239.63
Description & Specifications

Specifications

Chemical Name or Material FK 3311
CAS 116686-15-8
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C15H13F2NO4S
Formula Weight 341.33
Synonym N-[4-Acetyl-2-(2, 4-difluorophenoxy)phenyl]methanesulfonamide
Quantity 1mg
Safety and Handling

Store at Room Temperature