Please login to your online account to display your discounted pricing

FIT, Tocris

Irreversible δ agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material FIT
Assay Percent Range >99%
Molecular Formula C23H27N3OS
Formula Weight 393.55
Synonym N-[1-[2-(4-Isothiocyanatophenyl)ethyl]-4-piperidinyl]-N-phenylpropanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

148010

 
r & d systems
RDS148010
10mg Each for $239.63

148050

 
r & d systems
RDS148050
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 393.55
Quantity 10mg
Molecular Formula C23H27N3OS
Synonym N-[1-[2-(4-Isothiocyanatophenyl)ethyl]-4-piperidinyl]-N-phenylpropanamide
Chemical Name or Material FIT
Assay Percent Range >99%
CAS 85951-63-9
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C