Fisherbrand™ General Protection Disposable SMS Cranberry Lab Coats
Single-use lab coats provide high durability and splash/dust protection
$18.46 - $19.22
Specifications
Collar Type Standard
Cuff Style Knit
Closure Type Snap
No. of Pockets 3
Latex Free Yes
View More Specs
Products 7
Catalog Number Mfr. No. Size Length (English) Price Quantity  
Catalog Number Mfr. No. Size Length (English) Price Quantity  
17-100-857A View Documents Fisherbrand™
17100857A
Small 35 in. Pack of 10 Each for $18.46
Case of 3 Pack for $55.38
17-100-858A View Documents Fisherbrand™
17100858A
Medium 38 in. Pack of 10 Each for $18.85
Case of 3 Pack for $56.55
17-100-859A View Documents Fisherbrand™
17100859A
Large 39 in. Pack of 10 Each for $18.85
Case of 3 Pack for $56.55
17-100-860A View Documents Fisherbrand™
17100860A
X-Large 41 in. Pack of 10 Each for $18.85
Case of 3 Pack for $56.55
17-100-861A View Documents Fisherbrand™
17100861A
2X-Large 43 in. Pack of 10 Each for $19.22
Case of 3 Pack for $57.66
17-100-862A View Documents Fisherbrand™
17100862A
3X-Large 45 in. Pack of 10 Each for $19.22
Case of 3 Pack for $57.66
17-100-863A View Documents Fisherbrand™
17100863A
5X-Large 50 in. Pack of 10 Each for $19.22
Case of 3 Pack for $57.66
Description
Description
Roomy pockets for securly storing items like pens or small instruments
Latex-free, three-layer SMS construction
Packed 10 per pack; 30 per case
Excellent particle retention and water vapor transmission
Available in a variety of colors
Knit cuffs hold sleeves in place; standard collar
Three pockets and a five-snap closure
Fabric weight: 50g/m
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastic Cuffs, and Fisherbrand Basic Protection Disposable Lab Coats with Pockets. SMS coats are also available in White, Medical Blue, Teal, Purple and Blueberry.
Specifications
Specifications
Standard
Snap
Yes
SMS (Spunbond Meltblown Spunbond)
General SMS protection; Three pockets
Knit
3
Cranberry
Yes