Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
Supplier:  Fisherbrand™ C103
 View more versions of this product
Catalog No. 01-816E
Description
Description
Zipper-locking feature securely seals bags
  • Durable, transparent polyethylene
  • Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
  • Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Specifications
Not Autoclavable
Zip Seal
LDPE
50
13 x 9 in., (33 x 23cm)
33 cm
23 cm
3.25 lb.
Clear
250
4 mil
13 in.
9 in.