missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Y-Shaped Connectors with Straight Ends

Autoclavable and highly resistant to many chemicals.

Manufacturer:  Fisherbrand™ 222565

 View more versions of this product

Catalog No. 15-315-30C



Specifications

Specifications

Transparent
0.313 in.
Polypropylene
50.8mm
8mm
7.86mm
Y
1.27 in.
Yes
7.95mm
2 in.
Tubing Connector
20psi
0.31 in.
32.2mm
Documents