missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Y-Shaped Connectors with Straight Ends

Autoclavable and highly resistant to many chemicals.

Manufacturer:  Fisherbrand™ 222555

 View more versions of this product

Catalog No. 15-315-30B



Specifications

Specifications

Transparent
0.219 in.
Polypropylene
38.1mm
6mm
5.33mm
Y
0.8 in.
Yes
5.6mm
1.5 in.
Tubing Connector
20psi
0.21 in.
20.3mm
Documents