missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Y-Shaped Connectors with Straight Ends

Autoclavable and highly resistant to many chemicals.

Manufacturer:  Fisherbrand™ 222545

 View more versions of this product

Catalog No. 15-315-30A



Specifications

Specifications

Transparent
0.156 in.
Polypropylene
25.4mm
4mm
4mm
Y
0.56 in.
Yes
3.97mm
1 in.
Tubing Connector
20psi
0.16 in.
14.2mm
Documents