missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Y-Shaped Connectors with Straight Ends

Autoclavable and highly resistant to many chemicals.

Manufacturer:  Fisherbrand™ 222595

 View more versions of this product

Catalog No. 15-315-30F



Specifications

Specifications

Transparent
0.563 in.
Polypropylene
89mm
14mm
13.7mm
Y
2.1 in.
Yes
14.3mm
3.5 in.
Tubing Connector
20psi
0.54 in.
53.3mm
Documents