missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Y-Shaped Connectors with Straight Ends

Autoclavable and highly resistant to many chemicals.

Manufacturer:  Fisherbrand™ 222585

 View more versions of this product

Catalog No. 15-315-30E



Specifications

Specifications

Transparent
0.438 in.
Polypropylene
76.2mm
12mm
10.92mm
Y
1.78 in.
Yes
11.1mm
3 in.
Tubing Connector
20psi
0.43 in.
45.2mm
Documents