missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Wide-Mouth Field Sample Bottles

Bulk–packaged for convenient liquid sampling, transport and storage

Manufacturer:  Fisherbrand™ N3001410016

 View more versions of this product

Catalog No. 02-895-1C


 

Includes: Polypropylene screw caps

Description

Description

  • Excellent chemical resistance
  • Rigid and translucent

Specifications

Specifications

17 oz., 500mL
17 oz.
Translucent
53mm
Screw
500mL
Polypropylene
HDPE
53mm
Documents