missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Wide-Form Porcelain Crucibles

Withstand temperatures up to 1150°C (2102°F)

Manufacturer:  Fisherbrand™ J141

 View more versions of this product

Catalog No. FB965P



 

Alert:

Heating and cooling rates should not exceed 200°C per hour and maximum use temp of 1100°C


Specifications

Description

Crucible, Porcelain; Fisherbrand; Silico; High form; w/o Cover; Top O.D x Height x Wall Thickness: 102 x 60 x 2.3mm; 124g; Case wt.: 7.600kg; Case size: 490L x 340B x 315 in. H; Capac.: 250mL; 6/Pk.; 8 Pk./Cs.

  • Glazed inside and out except for outside bottom surface and rim

Description & Specifications

Specifications

8.45 oz.
8.45 oz., 250mL
102mm
2.36 in.
Porcelain
1150°C
2.3mm
124g
250mL
4.01 in.
Glazed
60mm
2102°F
0.09 in.
0.27 lb.
Crucible
Accessories